Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-tert-Butyl-3-thiosemicarbazide, 97%

$262.18 - $262.18

Chemical Identifiers

Chemical Name or Material 4-tert-Butyl-3-thiosemicarbazide
CAS 13431-39-5
Molecular Formula C5H13N3S
Formula Weight 147.24
MDL Number MFCD00041304
Beilstein 507219
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1070406

  SDS
alfa aesar™
L1070406
5g Each for $262.18
Description & Specifications

Specifications

Formula Weight 147.24
CAS 13431-39-5
Chemical Name or Material 4-tert-Butyl-3-thiosemicarbazide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Assay 97%
MDL Number MFCD00041304
Molecular Formula C5H13N3S
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 139° to 140°C
Beilstein 507219
Quantity 5g

Applications
Reatant in the synthesis of antiproliferative activity of N-substituted 2-amino-5-(2, 4-dihydroxyphenyl)-1, 3, 4-thiadiazoles.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.