Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-tert-Butyl-2-methylthiazole, 98%

$96.51 - $96.51

Chemical Identifiers

Chemical Name or Material 4-tert-Butyl-2-methylthiazole
CAS 15679-11-5
Molecular Formula C8H13NS
Formula Weight 155.26
MDL Number MFCD00051643
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1414806

 
alfa aesar™
A1414806
5g Each for $96.51
Description & Specifications

Specifications

Formula Weight 155.26
Chemical Name or Material 4-tert-Butyl-2-methylthiazole
Assay 98%
Molecular Formula C8H13NS
MDL Number MFCD00051643
CAS 15679-11-5
Boiling Point 106° to 108°C (70mmHg)
Odor Characteristic
Quantity 5g