Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-tert-Butyl-2-chlorophenol, tech. 90%

$28.93 - $263.22

Chemical Identifiers

Chemical Name or Material 4-tert-Butyl-2-chlorophenol
CAS 98-28-2
Purity Grade Technical Grade
Molecular Formula C10H13ClO
Formula Weight 184.66
MDL Number MFCD00019992
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2569514

  SDS
alfa aesar™
B2569514
25g Each for $28.93

AAB2569522

  SDS
alfa aesar™
B2569522
100g Each for $64.80

AAB2569536

  SDS
alfa aesar™
B2569536
500g Each for $263.22
Description & Specifications

Specifications

Formula Weight 184.66
Chemical Name or Material 4-tert-Butyl-2-chlorophenol
TSCA TSCA
Purity Grade Technical Grade
Assay 90%
Quantity 25g
Molecular Formula C10H13ClO
MDL Number MFCD00019992
CAS 98-28-2
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.