Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-tert-Butyl-2-chloroacetophenone, 97%

$102.05 - $380.20

Chemical Identifiers

Chemical Name or Material 4'-tert-Butyl-2-chloroacetophenone
CAS 21886-62-4
Molecular Formula C12H15ClO
Formula Weight 210.71
MDL Number MFCD00171621
Beilstein 1211242
Synonym alpha-Chloro-4-tert-butylacetophenone; 4-tert-Butylphenacyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2015603

  SDS
alfa aesar™
L2015603
1g Each for $102.05

AAL2015606

  SDS
alfa aesar™
L2015606
5g Each for $380.20
Description & Specifications

Specifications

Formula Weight 210.71
Chemical Name or Material 4'-tert-Butyl-2-chloroacetophenone
MDL Number MFCD00171621
CAS 21886-62-4
Synonym alpha-Chloro-4-tert-butylacetophenone; 4-tert-Butylphenacyl chloride
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Molecular Formula C12H15ClO
Beilstein 1211242
Health Hazard 1 Warning
Boiling Point 142° to 146°C (1mmHg)
Melting Point 36°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.