Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-tert-Butyl-2,6-diaminoanisole, 97%

Chemical Identifiers

Chemical Name or Material 4-tert-Butyl-2,6-diaminoanisole
CAS 473269-70-4
Formula Weight 194.28
MDL Number MFCD03701559
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5587803

 
alfa aesar™
H5587803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5587806

 
alfa aesar™
H5587806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5587814

 
alfa aesar™
H5587814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 194.28
Melting Point 111° to 113°C
Chemical Name or Material 4-tert-Butyl-2,6-diaminoanisole
Physical Form Powder
Assay 97%
MDL Number MFCD03701559
CAS 473269-70-4
Color White to orange
Quantity 1g