missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-[(tert-Butoxycarbonylamino)methyl]pyridine, TCI America™

$192.50 - $568.34

Chemical Identifiers

CAS 111080-65-0
Molecular Formula C11H16N2O2
Molecular Weight (g/mol) 208.261
MDL Number MFCD01321573
InChI Key KXCFJDZVQLRYSI-UHFFFAOYSA-N
Synonym tert-butyl pyridin-4-ylmethyl carbamate, n-boc-4-aminomethylpyridine, 4-tert-butoxycarbonylamino methyl pyridine, tert-butyl n-pyridin-4-ylmethyl carbamate, pyridin-4-ylmethyl carbamic acid tert-butyl ester, n-boc-4-aminomethylpyridine, tert-butyl pyridin-4-ylmethylcarbamate, 4-t-butyloxycarbonylaminomethyl-pyridine, 4-tert-butyloxycarbonylaminomethyl pyridin, 4-tert-butyloxycarbonylaminomethyl pyridine
PubChem CID 1512532
IUPAC Name tert-butyl N-(pyridin-4-ylmethyl)carbamate
SMILES CC(C)(C)OC(=O)NCC1=CC=NC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B21265G View Documents TCI America
B21265G
5g Each for $192.50
 
B212625G View Documents TCI America
B212625G
25g Each for $568.34
 
Specifications

Chemical Identifiers

111080-65-0
208.261
KXCFJDZVQLRYSI-UHFFFAOYSA-N
1512532
CC(C)(C)OC(=O)NCC1=CC=NC=C1
C11H16N2O2
MFCD01321573
tert-butyl pyridin-4-ylmethyl carbamate, n-boc-4-aminomethylpyridine, 4-tert-butoxycarbonylamino methyl pyridine, tert-butyl n-pyridin-4-ylmethyl carbamate, pyridin-4-ylmethyl carbamic acid tert-butyl ester, n-boc-4-aminomethylpyridine, tert-butyl pyridin-4-ylmethylcarbamate, 4-t-butyloxycarbonylaminomethyl-pyridine, 4-tert-butyloxycarbonylaminomethyl pyridin, 4-tert-butyloxycarbonylaminomethyl pyridine
tert-butyl N-(pyridin-4-ylmethyl)carbamate

Specifications

White-Yellow
5g
C11H16N2O2
tert-butyl pyridin-4-ylmethyl carbamate, n-boc-4-aminomethylpyridine, 4-tert-butoxycarbonylamino methyl pyridine, tert-butyl n-pyridin-4-ylmethyl carbamate, pyridin-4-ylmethyl carbamic acid tert-butyl ester, n-boc-4-aminomethylpyridine, tert-butyl pyridin-4-ylmethylcarbamate, 4-t-butyloxycarbonylaminomethyl-pyridine, 4-tert-butyloxycarbonylaminomethyl pyridin, 4-tert-butyloxycarbonylaminomethyl pyridine
CC(C)(C)OC(=O)NCC1=CC=NC=C1
208.261
208.26
≥98.0% (GC,T)
91°C
111080-65-0
MFCD01321573
KXCFJDZVQLRYSI-UHFFFAOYSA-N
tert-butyl N-(pyridin-4-ylmethyl)carbamate
1512532
Crystal-Powder at 20°C
4-[(tert-Butoxycarbonylamino)methyl]pyridine
Safety and Handling

Safety and Handling

TSCA : No