missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-[(tert-Butoxycarbonylamino)methyl]pyridine-2-carboxylic Acid, TCI America™

[Reagent for application of the exciton chirality method]

$239.86 - $1,287.94

Chemical Identifiers

CAS 260970-57-8
Molecular Formula C12H16N2O4
Molecular Weight (g/mol) 252.27
MDL Number MFCD02093452
InChI Key BSNYTBOSHJFWQM-UHFFFAOYSA-N
Synonym 4-(Boc-aminomethyl)pyridine-2-carboxylic Acid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B21241G View Documents TCI America
B21241G
1g Each for $1,287.94
 
B2124100MG View Documents TCI America
B2124100MG
100mg Each for $239.86
 
Specifications

Chemical Identifiers

260970-57-8
252.27
BSNYTBOSHJFWQM-UHFFFAOYSA-N
C12H16N2O4
MFCD02093452
4-(Boc-aminomethyl)pyridine-2-carboxylic Acid

Specifications

White-Yellow
260970-57-8
MFCD02093452
BSNYTBOSHJFWQM-UHFFFAOYSA-N
252.27
≥96.0% (HPLC,T)
1g
C12H16N2O4
4-(Boc-aminomethyl)pyridine-2-carboxylic Acid
252.27
Crystal-Powder at 20°C
4-[(tert-Butoxycarbonylamino)methyl]pyridine-2-carboxylic Acid [Reagent for application of the exciton chirality method]
Safety and Handling

Safety and Handling

TSCA : No