missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(tert-Butoxycarbonylamino)-1-butanol, TCI America™

$252.09 - $855.58

Chemical Identifiers

CAS 75178-87-9
Molecular Formula C9H19NO3
Molecular Weight (g/mol) 189.255
InChI Key LIYMTLVBAVHPBU-UHFFFAOYSA-N
Synonym tert-butyl 4-hydroxybutyl carbamate, tert-butyl n-4-hydroxybutyl carbamate, 4-boc-amino-1-butanol, 4-tert-butoxycarbonylamino-1-butanol, carbamic acid, 4-hydroxybutyl-, 1,1-dimethylethyl ester, tert-butyl 4-hydroxybutylcarbamate, n-boc-4-aminobutanol, butylhydroxybutylcarbamate, acmc-1bfe1, 4-t-boc-amino-1-butanol
PubChem CID 545141
IUPAC Name tert-butyl N-(4-hydroxybutyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B28685G View Documents TCI America
B28685G
5g Each for $252.09
 
B286825G View Documents TCI America
B286825G
25g Each for $855.58
 
Specifications

Chemical Identifiers

75178-87-9
189.255
tert-butyl 4-hydroxybutyl carbamate, tert-butyl n-4-hydroxybutyl carbamate, 4-boc-amino-1-butanol, 4-tert-butoxycarbonylamino-1-butanol, carbamic acid, 4-hydroxybutyl-, 1,1-dimethylethyl ester, tert-butyl 4-hydroxybutylcarbamate, n-boc-4-aminobutanol, butylhydroxybutylcarbamate, acmc-1bfe1, 4-t-boc-amino-1-butanol
tert-butyl N-(4-hydroxybutyl)carbamate
C9H19NO3
LIYMTLVBAVHPBU-UHFFFAOYSA-N
545141
CC(C)(C)OC(=O)NCCCCO

Specifications

White-Yellow
75178-87-9
tert-butyl 4-hydroxybutyl carbamate, tert-butyl n-4-hydroxybutyl carbamate, 4-boc-amino-1-butanol, 4-tert-butoxycarbonylamino-1-butanol, carbamic acid, 4-hydroxybutyl-, 1,1-dimethylethyl ester, tert-butyl 4-hydroxybutylcarbamate, n-boc-4-aminobutanol, butylhydroxybutylcarbamate, acmc-1bfe1, 4-t-boc-amino-1-butanol
CC(C)(C)OC(=O)NCCCCO
189.255
189.26
≥96.0% (GC)
5g
C9H19NO3
LIYMTLVBAVHPBU-UHFFFAOYSA-N
tert-butyl N-(4-hydroxybutyl)carbamate
545141
Crystal-Lump at 20°C
4-(tert-Butoxycarbonylamino)-1-butanol
Safety and Handling

Safety and Handling

TSCA : No