missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(tert-Butoxycarbonyl)benzeneboronic acid, 95%

Manufacturer:  Alfa Aesar™ H53198MD

 View more versions of this product

Catalog No. AAH53198MD



Description

Description

Applications
Benzene boronic acid mediated the ortho specific hydroxyalkylation of phenols by aldehydes. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. It can be a effective catalyst for amidation and esterification of carboxylic acids.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Specifications

Specifications

4-(tert-Butoxycarbonyl)benzeneboronic acid
250mg
C11H15BO4
4-tert-butoxycarbonyl phenylboronic acid, 4-tert-butoxycarbonyl phenyl boronic acid, 4-t-butoxycarbonyl phenylboronic acid, tert-butyl 4-boronobenzoate, 4-t-butoxycarbonylphenylboronic acid, 4-boc-phenylboronic acid, 4-tert-butoxycarbonyl benzeneboronic acid, 4-tert-butoxycarbonyl phenyl boronic acid, benzoic acid,4-borono-, 1,1-dimethylethyl ester, 4-t-butoxycarbonyl phenylboronicacid
B(C1=CC=C(C=C1)C(=O)OC(C)(C)C)(O)O
222.047
222.05
100°C to 106°C
850568-54-6
MFCD03411946
QMVMDYSTJSUDKC-UHFFFAOYSA-N
[4-[(2-methylpropan-2-yl)oxycarbonyl]phenyl]boronic acid
2773301
95%

Chemical Identifiers

850568-54-6
222.047
QMVMDYSTJSUDKC-UHFFFAOYSA-N
2773301
B(C1=CC=C(C=C1)C(=O)OC(C)(C)C)(O)O
C11H15BO4
MFCD03411946
4-tert-butoxycarbonyl phenylboronic acid, 4-tert-butoxycarbonyl phenyl boronic acid, 4-t-butoxycarbonyl phenylboronic acid, tert-butyl 4-boronobenzoate, 4-t-butoxycarbonylphenylboronic acid, 4-boc-phenylboronic acid, 4-tert-butoxycarbonyl benzeneboronic acid, 4-tert-butoxycarbonyl phenyl boronic acid, benzoic acid,4-borono-, 1,1-dimethylethyl ester, 4-t-butoxycarbonyl phenylboronicacid
[4-[(2-methylpropan-2-yl)oxycarbonyl]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents