Please login to your online account to display your discounted pricing

4-[(tert-Butoxycarbonyl)amino]-1-methyl-1H-pyrrole-2-carboxylic acid, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SPB08449CB

 View more versions of this product

Catalog No. SPB08449CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 240.26
Quantity 250mg
Assay 97%
CAS Min % 97.0
MDL Number MFCD02094544
Chemical Name or Material 4-[(tert-Butoxycarbonyl)amino]-1-methyl-1H-pyrrole-2-carboxylic acid
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H16N2O4
CAS 77716-11-1

Specifications

Chemical Name or Material 4-[(tert-Butoxycarbonyl)amino]-1-methyl-1H-pyrrole-2-carboxylic acid
CAS 77716-11-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H16N2O4
Formula Weight 240.26
MDL Number MFCD02094544