Please login to your online account to display your discounted pricing

4-[(tert-Butoxycarbonyl)amino]-1-methyl-1H-pyrrole-2-carboxylic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-[(tert-Butoxycarbonyl)amino]-1-methyl-1H-pyrrole-2-carboxylic acid
CAS 77716-11-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H16N2O4
Formula Weight 240.26
MDL Number MFCD02094544
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB08449CB

  SDS
fisher scientific company
SPB08449CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 240.26
Quantity 250mg
Assay 97%
CAS Min % 97.0
MDL Number MFCD02094544
Packaging Amber Glass Bottle
CAS 77716-11-1
Assay Percent Range 97%
Molecular Formula C11H16N2O4
Chemical Name or Material 4-[(tert-Butoxycarbonyl)amino]-1-methyl-1H-pyrrole-2-carboxylic acid