missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-tert-Butoxybenzonitrile, 96%, Thermo Scientific™
$118.62 - $404.00
Chemical Identifiers
CAS | 185259-36-3 |
---|---|
Molecular Formula | C11H13NO |
Molecular Weight (g/mol) | 175.231 |
MDL Number | MFCD03425273 |
InChI Key | WLWYCCYGVDZFMS-UHFFFAOYSA-N |
Synonym | 4-tert-butoxybenzonitrile, 4-tert-butoxy benzonitrile, benzonitrile, 4-1,1-dimethylethoxy, acmc-20amau, 4-tert-butoxy benzenecarbonitrile, 4-2-methylpropan-2-yl oxy benzonitrile |
PubChem CID | 7021978 |
IUPAC Name | 4-[(2-methylpropan-2-yl)oxy]benzonitrile |
SMILES | CC(C)(C)OC1=CC=C(C=C1)C#N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1806706
|
Thermo Scientific Chemicals
L1806706 |
5 g |
Each for $118.62
|
|
|||||
AAL1806714
|
Thermo Scientific Chemicals
L1806714 |
25 g |
Each for $404.00
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
185259-36-3 | |
175.231 | |
WLWYCCYGVDZFMS-UHFFFAOYSA-N | |
7021978 | |
CC(C)(C)OC1=CC=C(C=C1)C#N |
C11H13NO | |
MFCD03425273 | |
4-tert-butoxybenzonitrile, 4-tert-butoxy benzonitrile, benzonitrile, 4-1,1-dimethylethoxy, acmc-20amau, 4-tert-butoxy benzenecarbonitrile, 4-2-methylpropan-2-yl oxy benzonitrile | |
4-[(2-methylpropan-2-yl)oxy]benzonitrile |
Specifications
185259-36-3 | |
C11H13NO | |
MFCD03425273 | |
3237723 | |
WLWYCCYGVDZFMS-UHFFFAOYSA-N | |
4-[(2-methylpropan-2-yl)oxy]benzonitrile | |
7021978 | |
96% |
95°C (0.8 mmHg) | |
1.521 | |
5 g | |
4-tert-butoxybenzonitrile, 4-tert-butoxy benzonitrile, benzonitrile, 4-1,1-dimethylethoxy, acmc-20amau, 4-tert-butoxy benzenecarbonitrile, 4-2-methylpropan-2-yl oxy benzonitrile | |
CC(C)(C)OC1=CC=C(C=C1)C#N | |
175.231 | |
175.23 | |
4-tert-Butoxybenzonitrile |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only