Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-tert-Butoxybenzaldehyde, 98%

$195.39 - $195.39

Chemical Identifiers

Chemical Name or Material 4-tert-Butoxybenzaldehyde
CAS 57699-45-3
Molecular Formula C11H14O2
Formula Weight 178.23g/mol
MDL Number MFCD00040800
Beilstein 5499455
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1343603

 
alfa aesar™
L13436-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1343606

 
alfa aesar™
L13436-06
5g Each for $195.39
Description & Specifications

Specifications

Boiling Point 87° to 92°C (1.5mmHg)
Quantity 1g
Chemical Name or Material 4-tert-Butoxybenzaldehyde
CAS 57699-45-3
Assay 98%
Molecular Formula C11H14O2
Formula Weight 178.23g/mol
MDL Number MFCD00040800
Beilstein 5499455
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.