Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Sulfophthalic acid, tech., 50% w/w aq. soln.

Chemical Identifiers

Chemical Name or Material 4-Sulfophthalic acid
CAS 89-08-7
Purity Grade Technical Grade
Molecular Formula C8H6O7S
Formula Weight 246.2
MDL Number MFCD00007494
UN Number UN3265
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1631836

 
alfa aesar™
A1631836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 246.2
Health Hazard 2 GHS H Statement
H314
Causes severe skin burns and eye damage.
Chemical Name or Material 4-Sulfophthalic acid
Concentration 50% w/w aq. soln.
Health Hazard 1 Danger
Quantity 500g
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA TSCA
Molecular Formula C8H6O7S
CAS 89-08-7
Purity Grade Technical Grade
MDL Number MFCD00007494
UN Number UN3265
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314
Causes severe skin burns and eye damage.