Please login to your online account to display your discounted pricing

4-Sulfophthalic Acid, 50% in Water Pract., ACROS Organics™


Quantity 2kg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  419930020

 View more versions of this product

Catalog No. AC419930020

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Density 1.2900g/mL
Packaging Glass bottle
Refractive Index 1.4450 to 1.4490 (20°C, 589nm)
Color Clear slightly brown
Quantity 2kg
Chemical Name or Material 4-Sulfophthalic acid
Name Note pract.
CAS 89-08-7
CAS 7732-18-5
Physical Form Clear slightly brown liquid
Assay 46 to 54%
Molecular Formula C8H6O7S
Formula Weight 246.20g/mol
MDL Number MFCD00007494
Beilstein 11, 406
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 201-881-6
TSCA TSCA

Specifications

Chemical Name or Material 4-Sulfophthalic acid
Name Note pract.
CAS 89-08-7
CAS 7732-18-5
Assay 46 to 54%
Molecular Formula C8H6O7S
Formula Weight 246.20g/mol
MDL Number MFCD00007494
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents