Please login to your online account to display your discounted pricing

4-Sulfophthalic Acid, 50% in Water Pract., ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Sulfophthalic acid
Name Note pract.
CAS 89-08-7
CAS 7732-18-5
Assay 46 to 54%
Molecular Formula C8H6O7S
Formula Weight 246.2
MDL Number MFCD00007494
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC419930020

  SDS
thermo fisher scientific
AC419930020
Glass bottle 2kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC419930250

  SDS
thermo fisher scientific
419930250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 246.2
Quantity 2kg
Color Clear slightly brown
EINECS Number 201-881-6
TSCA TSCA
Density 1.2900g/mL
Assay 46 to 54%
Beilstein 11, 406
Health Hazard 1 Danger
Packaging Glass bottle
Refractive Index 1.4450 to 1.4490 (20°C, 589nm)
Chemical Name or Material 4-Sulfophthalic acid
Name Note pract.
Molecular Formula C8H6O7S
CAS 89-08-7
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
CAS 7732-18-5
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00007494
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.