Please login to your online account to display your discounted pricing

4-Sulfophthalic Acid, 50% in Water Pract., ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  419930250

 View more versions of this product

Catalog No. AC419930250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 246.2
Physical Form Liquid
Quantity 25g
Color Clear slightly brown
Beilstein 11, 406
Assay 46 to 54%
TSCA TSCA
EINECS Number 201-881-6
Density 1.2900g/mL
Health Hazard 1 Danger
CAS 7732-18-5
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.4450 to 1.4490 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Molecular Formula C8H6O7S
Chemical Name or Material 4-Sulfophthalic acid
Name Note pract.
MDL Number MFCD00007494
CAS 89-08-7

Specifications

Chemical Name or Material 4-Sulfophthalic acid
Name Note pract.
CAS 89-08-7
CAS 7732-18-5
Assay 46 to 54%
Molecular Formula C8H6O7S
Formula Weight 246.2
MDL Number MFCD00007494
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents