missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Sulfonic-calix[6]arene hydrate, Alfa Aesar™

Manufacturer:  Alfa Aesar L1362503

 View more versions of this product

Catalog No. AAL1362503



Specifications

Specifications

4-Sulfonic-calix[6]arene hydrate
C42H36O24S6
5231405
BXOUPTJVBGEDIR-UHFFFAOYSA-N
1117.086
1117.12 (Anhydrous)
1g
102088-39-1
MFCD00216907
4-sulfocalix 6 arene, 4-sulfocalix 6 arene hydrate, hexahydroxycalix 6 arenehexasulfonic acid, 4-sulfonic calix 6 arene, acmc-20m53e, calix 6 arene-p-sulfonate, hexasulfo hexahydroxy calix 6 arene
C1C2=CC(=CC(=C2O)CC3=CC(=CC(=C3O)CC4=CC(=CC(=C4O)CC5=C(C(=CC(=C5)S(=O)(=O)O)CC6=C(C(=CC(=C6)S(=O)(=O)O)CC7=C(C1=CC(=C7)S(=O)(=O)O)O)O)O)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O
2724887
~246°C

Chemical Identifiers

102088-39-1
1117.086
BXOUPTJVBGEDIR-UHFFFAOYSA-N
2724887
C42H36O24S6
MFCD00216907
4-sulfocalix 6 arene, 4-sulfocalix 6 arene hydrate, hexahydroxycalix 6 arenehexasulfonic acid, 4-sulfonic calix 6 arene, acmc-20m53e, calix 6 arene-p-sulfonate, hexasulfo hexahydroxy calix 6 arene
C1C2=CC(=CC(=C2O)CC3=CC(=CC(=C3O)CC4=CC(=CC(=C4O)CC5=C(C(=CC(=C5)S(=O)(=O)O)CC6=C(C(=CC(=C6)S(=O)(=O)O)CC7=C(C1=CC(=C7)S(=O)(=O)O)O)O)O)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents