missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Sulfonic calix[6]arene hydrate, 95%, ACROS Organics™

Chemical Identifiers

CAS 205652-55-7
Molecular Formula C42H38O25S6
Molecular Weight (g/mol) 1135.101
MDL Number MFCD00216907
InChI Key SWWAZWAJPNREHZ-UHFFFAOYSA-N
Synonym 4-sulfonic-calix 6 arene hydrate
PubChem CID 121233553
SMILES C1C2=CC(=CC(=C2O)CC3=CC(=CC(=C3O)CC4=CC(=CC(=C4O)CC5=C(C(=CC(=C5)S(=O)(=O)O)CC6=C(C(=CC(=C6)S(=O)(=O)O)CC7=C(C1=CC(=C7)S(=O)(=O)O)O)O)O)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O.O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC290260010 View Documents Acros Organics
290260010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Chelation/Complexation reagent
Specifications

Chemical Identifiers

205652-55-7
1135.101
SWWAZWAJPNREHZ-UHFFFAOYSA-N
121233553
C42H38O25S6
MFCD00216907
4-sulfonic-calix 6 arene hydrate
C1C2=CC(=CC(=C2O)CC3=CC(=CC(=C3O)CC4=CC(=CC(=C4O)CC5=C(C(=CC(=C5)S(=O)(=O)O)CC6=C(C(=CC(=C6)S(=O)(=O)O)CC7=C(C1=CC(=C7)S(=O)(=O)O)O)O)O)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O.O

Specifications

Authentic
Beige to Brown-Gray
205652-55-7
100.0
C42H38O25S6
4-sulfonic-calix 6 arene hydrate
C1C2=CC(=CC(=C2O)CC3=CC(=CC(=C3O)CC4=CC(=CC(=C4O)CC5=C(C(=CC(=C5)S(=O)(=O)O)CC6=C(C(=CC(=C6)S(=O)(=O)O)CC7=C(C1=CC(=C7)S(=O)(=O)O)O)O)O)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O.O
121233553
Powder
4-Sulfonic calix[6]arene hydrate
Glass bottle
1g
94.0
95%
MFCD00216907
SWWAZWAJPNREHZ-UHFFFAOYSA-N
1135.101
1117.13
≥94%