Please login to your online account to display your discounted pricing

4-Sulfonic calix[6]arene hydrate, 95%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  290260010

 View more versions of this product

Catalog No. AC290260010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 1117.13
Physical Form Powder
Assay ≥94%
Quantity 1g
Color Light beige to gray-brown
CAS Max % 100.0
CAS Min % 94.0
Packaging Glass bottle
MDL Number MFCD00216907
Assay Percent Range 95%
Chemical Name or Material 4-Sulfonic calix[6]arene hydrate
Molecular Formula C42H36O24S6·xH2O
CAS 205652-55-7
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4-Sulfonic calix[6]arene hydrate
CAS 205652-55-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C42H36O24S6·xH2O
Formula Weight 1117.13
MDL Number MFCD00216907