Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Sulfonic-calix[6]arene hydrate

$456.55 - $456.55

Chemical Identifiers

Chemical Name or Material 4-Sulfonic-calix[6]arene hydrate
CAS 102088-39-1
Molecular Formula C42H36O24S6·xH2O
Formula Weight 1117.12 (Anhydrous)
MDL Number MFCD00216907
Beilstein 5231405
Synonym Calix[6]arene-4-sulfonic acid hydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1362506

  SDS
alfa aesar™
L1362506
5g Each for $456.55
Description & Specifications

Specifications

Formula Weight 1117.12 (Anhydrous)
Chemical Name or Material 4-Sulfonic-calix[6]arene hydrate
Synonym Calix[6]arene-4-sulfonic acid hydrate
CAS 102088-39-1
Molecular Formula C42H36O24S6·xH2O
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 5231405
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈246°C
Quantity 5g
MDL Number MFCD00216907
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.