missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Sulfonic calix[4]arene hydrate, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC290250050
Description
Chelation/Complexation reagentSpecifications
4-Sulfonic calix[4]arene hydrate | |
Glass bottle | |
5g | |
C28H24O16S4·xH2O | |
Solubility in water: soluble. Other solubilities: soluble in methanol and ethanol | |
744.75 | |
95% |
Authentic | |
Beige-Gray to White | |
94% min. (HPLC) | |
MFCD16495912 | |
744.75 | |
Powder |
Chemical Identifiers
C28H24O16S4·xH2O | |
744.75 | |
MFCD16495912 |