Please login to your online account to display your discounted pricing

4-Sulfobenzoic acid monopotassium salt, 98%, ACROS Organics™

$34.18 - $34.18

Chemical Identifiers

Chemical Name or Material 4-Sulfobenzoic acid monopotassium salt
CAS 5399-63-3
CAS Min % 97.5
CAS Max % 100.0
Purity Grade Practical Grade
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H5KO5S
Linear Formula SO3KC6H4CO2H
Formula Weight 240.28
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC419880250

  SDS
thermo fisher scientific
419880250
Glass bottle 25g Each for $34.18
Description & Specifications

Specifications

Assay ≥97.5%
Purity Grade Practical Grade
Physical Form Fine Crystalline Powder
Formula Weight 240.28
Quantity 25g
Color White
CAS Max % 100.0
Linear Formula SO3KC6H4CO2H
CAS 5399-63-3
Chemical Name or Material 4-Sulfobenzoic acid monopotassium salt
MDL Number MFCD00007509
CAS Min % 97.5
Packaging Glass bottle
Molecular Formula C7H5KO5S
Infrared Spectrum Authentic
Assay Percent Range 98%