Please login to your online account to display your discounted pricing

4-Sulfobenzoic acid monopotassium salt, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  419880250

 View more versions of this product

Catalog No. AC419880250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Purity Grade Practical Grade
Physical Form Fine Crystalline Powder
Formula Weight 240.28
Quantity 25g
Color White
CAS Max % 100.0
Linear Formula SO3KC6H4CO2H
CAS 5399-63-3
Chemical Name or Material 4-Sulfobenzoic acid monopotassium salt
MDL Number MFCD00007509
CAS Min % 97.5
Packaging Glass bottle
Molecular Formula C7H5KO5S
Infrared Spectrum Authentic
Assay Percent Range 98%

Specifications

Chemical Name or Material 4-Sulfobenzoic acid monopotassium salt
CAS 5399-63-3
CAS Min % 97.5
CAS Max % 100.0
Purity Grade Practical Grade
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H5KO5S
Linear Formula SO3KC6H4CO2H
Formula Weight 240.28
MDL Number MFCD00007509