Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Sulfamoylbenzoic acid, 96%

$58.35 - $58.35

Chemical Identifiers

Chemical Name or Material 4-Sulfamoylbenzoic acid
CAS 138-41-0
Molecular Formula C7H7NO4S
Formula Weight 201.2
MDL Number MFCD00007938
Beilstein 1875393
Merck Index 14,1876
Synonym 4-(Aminosulfonyl)benzoic acid; 4-Carboxybenzenesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2244918

 
alfa aesar™
B2244918
50g Each for $58.35
Description & Specifications

Specifications

Formula Weight 201.2
Chemical Name or Material 4-Sulfamoylbenzoic acid
TSCA TSCA
Assay 96%
Molecular Formula C7H7NO4S
MDL Number MFCD00007938
CAS 138-41-0
Beilstein 1875393
Merck Index 14,1876
Synonym 4-(Aminosulfonyl)benzoic acid; 4-Carboxybenzenesulfonamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈285°C (dec.)
RTECS Number DH6820000
Quantity 50g

Applications
Sulfamoylbenzoic acid is a moiety of sulfamyl diuretics or antihypertensive. It is used as an antiviral and an intermediate for the manufacture of frusemide which used as a diuretic and as an antihypertensive.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.