missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-sec-Butylphenol, TCI America™

$41.25 - $339.17

Chemical Identifiers

CAS 99-71-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00002375
InChI Key ZUTYZAFDFLLILI-UHFFFAOYSA-N
PubChem CID 7453
ChEBI CHEBI:34442
IUPAC Name 4-butan-2-ylphenol
SMILES CCC(C)C1=CC=C(C=C1)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B073325G View Documents TCI America
B073325G
25g Each for $41.25
 
B0733100G View Documents TCI America
B0733100G
100g Each for $105.42
 
B0733500G View Documents TCI America
B0733500G
500g Each for $339.17
 
Specifications

Chemical Identifiers

99-71-8
150.221
ZUTYZAFDFLLILI-UHFFFAOYSA-N
CHEBI:34442
CCC(C)C1=CC=C(C=C1)O
C10H14O
MFCD00002375
7453
4-butan-2-ylphenol

Specifications

110°C
60°C
99-71-8
MFCD00002375
ZUTYZAFDFLLILI-UHFFFAOYSA-N
4-butan-2-ylphenol
7453
150.22
≥98.0% (GC)
White
25g
C10H14O
2430
CCC(C)C1=CC=C(C=C1)O
150.221
CHEBI:34442
Crystal-Powder at 20°C
4-sec-Butylphenol
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0503

RTECSNumber : SJ8924000

TSCA : Yes