missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-sec-Butylbenzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H53358MD

 View more versions of this product

Catalog No. AAH53358MD



Specifications

Specifications

4-sec-Butylbenzeneboronic acid
250mg
C10H15BO2
4-sec-butylphenylboronic acid, 4-sec-butylphenyl boronic acid, 4-1-methylpropyl benzeneboronic acid, 4-sec-butyl phenyl boronic acid, 4-1-methylpropyl phenylboronic acid, 4-sec-butyl benzeneboronic acid, 4-sec-butyl phenylboronic acid, 4-butan-2-yl phenyl boronic acid, boronic acid,b-4-1-methylpropyl phenyl, boronic acid, 4-1-methylpropyl phenyl
B(C1=CC=C(C=C1)C(C)CC)(O)O
178.038
178.04
96°C to 100°C
850568-56-8
MFCD02179456
CHSPURIZKQPJNH-UHFFFAOYSA-N
(4-butan-2-ylphenyl)boronic acid
3690874
98%

Chemical Identifiers

850568-56-8
178.038
CHSPURIZKQPJNH-UHFFFAOYSA-N
3690874
B(C1=CC=C(C=C1)C(C)CC)(O)O
C10H15BO2
MFCD02179456
4-sec-butylphenylboronic acid, 4-sec-butylphenyl boronic acid, 4-1-methylpropyl benzeneboronic acid, 4-sec-butyl phenyl boronic acid, 4-1-methylpropyl phenylboronic acid, 4-sec-butyl benzeneboronic acid, 4-sec-butyl phenylboronic acid, 4-butan-2-yl phenyl boronic acid, boronic acid,b-4-1-methylpropyl phenyl, boronic acid, 4-1-methylpropyl phenyl
(4-butan-2-ylphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents