missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Quinolinol 98.0+%, TCI America™

Manufacturer:  TCI America Q00295G

 View more versions of this product

Catalog No. Q00295G



Specifications

Specifications

4-Quinolinol
203°C
611-36-9
MFCD00006777
PMZDQRJGMBOQBF-UHFFFAOYSA-N
1H-quinolin-4-one
69141
145.16
≥98.0% (GC,T)
White-Yellow
5g
C9H7NO
4-hydroxyquinoline, quinolin-4-ol, 4-quinolinol, quinolin-4 1h-one, 4 1h-quinolinone, kynurine, 4-hydroxy quinoline, quinoline, 4-hydroxy, ccris 4329, 4-chinolinol
C1=CC=C2C(=C1)C(=O)C=CN2
145.161
CHEBI:15815
Crystal-Powder at 20°C

Chemical Identifiers

611-36-9
145.161
PMZDQRJGMBOQBF-UHFFFAOYSA-N
69141
1H-quinolin-4-one
C9H7NO
MFCD00006777
4-hydroxyquinoline, quinolin-4-ol, 4-quinolinol, quinolin-4 1h-one, 4 1h-quinolinone, kynurine, 4-hydroxy quinoline, quinoline, 4-hydroxy, ccris 4329, 4-chinolinol
CHEBI:15815
C1=CC=C2C(=C1)C(=O)C=CN2
Safety and Handling

Safety and Handling

RTECSNumber : VC4070000

TSCA : No

Documents