Please login to your online account to display your discounted pricing

4-(Pyrrolidine-1-sulfonyl)-benzenesulfonyl chloride, Technical Grade, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(Pyrrolidine-1-sulfonyl)-benzenesulfonyl chloride
Assay Tech.
Molecular Formula C10H12ClNO4S2
Formula Weight 309.79
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00517CB

  SDS
fisher scientific company
MO00517CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 309.79
Purity Grade Technical Grade
Quantity 250mg
Assay Tech.
CAS 165669-32-9
Chemical Name or Material 4-(Pyrrolidine-1-sulfonyl)-benzenesulfonyl chloride
Molecular Formula C10H12ClNO4S2