Please login to your online account to display your discounted pricing

4-(Pyrrolidine-1-sulfonyl)-benzenesulfonyl chloride, Technical Grade, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  MO00517CB

 View more versions of this product

Catalog No. MO00517CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 309.79
Quantity 250mg
Assay Tech.
Chemical Name or Material 4-(Pyrrolidine-1-sulfonyl)-benzenesulfonyl chloride
Molecular Formula C10H12ClNO4S2
CAS 165669-32-9

Specifications

Chemical Name or Material 4-(Pyrrolidine-1-sulfonyl)-benzenesulfonyl chloride
Assay Tech.
Molecular Formula C10H12ClNO4S2
Formula Weight 309.79