Please login to your online account to display your discounted pricing

4-(Pyrrolidin-1-ylmethyl)benzoic acid, 97%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  CC55601CB

 View more versions of this product

Catalog No. CC55601CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 205.26
Quantity 250mg
Assay 97%
Chemical Name or Material 4-(Pyrrolidin-1-ylmethyl)benzoic acid
Molecular Formula C12H15NO2
CAS 159691-25-5
Assay Percent Range 97%

Specifications

Chemical Name or Material 4-(Pyrrolidin-1-ylmethyl)benzoic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H15NO2
Formula Weight 205.26