Please login to your online account to display your discounted pricing

4-(Pyrrolidin-1-ylmethyl)aniline, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(Pyrrolidin-1-ylmethyl)aniline
Assay Percent Range 97%
Molecular Formula C11H16N2
Formula Weight 176.26
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

CC55614EA

 
fisher scientific company
CC55614EA
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC55614DA

 
fisher scientific company
CC55614DA
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC55614CB

 
fisher scientific company
CC55614CB
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 176.26
Quantity 10g
Chemical Name or Material 4-(Pyrrolidin-1-ylmethyl)aniline
Assay Percent Range 97%
Molecular Formula C11H16N2
CAS 142335-64-6