Please login to your online account to display your discounted pricing

4-(Pyrrolidin-1-ylmethyl)aniline, 97%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  CC55614CB

 View more versions of this product

Catalog No. CC55614CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 176.26
Quantity 250mg
CAS 142335-64-6
Chemical Name or Material 4-(Pyrrolidin-1-ylmethyl)aniline
Assay Percent Range 97%
Molecular Formula C11H16N2

Specifications

Chemical Name or Material 4-(Pyrrolidin-1-ylmethyl)aniline
Assay Percent Range 97%
Molecular Formula C11H16N2
Formula Weight 176.26