Please login to your online account to display your discounted pricing

4-Pyrimidinecarboxaldehyde, 95%, Acros Organics


Quantity 250mg

Manufacturer: thermo fisher scientific  AC454292500

 View more versions of this product

Catalog No. AC454292500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 108.1
Quantity 250mg
Assay Percent Range 95%
Chemical Name or Material 4-Pyrimidinecarboxaldehyde
CAS 2435-50-9
Molecular Formula C5H4N2O

Specifications

Chemical Name or Material 4-Pyrimidinecarboxaldehyde
Assay Percent Range 95%
Molecular Formula C5H4N2O
Formula Weight 108.1