missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Pyrimidinecarboxaldehyde, 95%, ACROS Organics™

Chemical Identifiers

CAS 2435-50-9
Molecular Formula C5H4N2O
Molecular Weight (g/mol) 108.1
InChI Key OKULHRWWYCFJAB-UHFFFAOYSA-N
Synonym 4-formyl-1,3-diazine, 4-formylpyrimidine, 4-pyrimidine carbaldehyde, 4-pyrimidinecarboxaldehyde, 4-pyrimidinecarboxaldehyde 7ci,8ci,9ci, acmc-1cp8l, pyrimidine-4-carboxaldehyde
PubChem CID 11412355
IUPAC Name pyrimidine-4-carbaldehyde
SMILES C1=CN=CN=C1C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC454290010
SDS View Documents
Thermo Fisher Scientific
AC454290010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

2435-50-9
108.1
4-formyl-1,3-diazine, 4-formylpyrimidine, 4-pyrimidine carbaldehyde, 4-pyrimidinecarboxaldehyde, 4-pyrimidinecarboxaldehyde 7ci,8ci,9ci, acmc-1cp8l, pyrimidine-4-carboxaldehyde
pyrimidine-4-carbaldehyde
C5H4N2O
OKULHRWWYCFJAB-UHFFFAOYSA-N
11412355
C1=CN=CN=C1C=O

Specifications

Glass Bottle
2435-50-9
C5H4N2O
OKULHRWWYCFJAB-UHFFFAOYSA-N
pyrimidine-4-carbaldehyde
11412355
95%
1g
94% min. (GC)
4-formyl-1,3-diazine, 4-formylpyrimidine, 4-pyrimidine carbaldehyde, 4-pyrimidinecarboxaldehyde, 4-pyrimidinecarboxaldehyde 7ci,8ci,9ci, acmc-1cp8l, pyrimidine-4-carboxaldehyde
C1=CN=CN=C1C=O
108.1
108.1
4-Pyrimidinecarboxaldehyde
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS