missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Pyrimidin-5-ylbenzaldehyde, ≥95%, Maybridge

Chemical Identifiers

CAS 198084-12-7
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C11H8N2O
Formula Weight 184.2
MDL Number MFCD02684108
Chemical Name or Material 4-Pyrimidin-5-ylbenzaldehyde
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC44304DA SDS Fisher Scientific Company
CC44304DA
Amber glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

198084-12-7
100.0
C11H8N2O
MFCD02684108
95%
94.5
95%
184.2
4-Pyrimidin-5-ylbenzaldehyde

Specifications

184.2
100.0
198084-12-7
95%
C11H8N2O
95%
1g
MFCD02684108
94.5
Amber glass bottle
4-Pyrimidin-5-ylbenzaldehyde

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.