Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Pyridylthio)acetic acid, 98%

$36.32 - $395.59

Chemical Identifiers

Chemical Name or Material (4-Pyridylthio)acetic acid
CAS 10351-19-6
Molecular Formula C7H7NO2S
Formula Weight 169.2
MDL Number MFCD00006424
Beilstein 129738
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2149914

 
alfa aesar™
B2149914
25g Each for $36.32

AAB2149936

 
alfa aesar™
B2149936
500g Each for $395.59
Description & Specifications

Specifications

Formula Weight 169.2
Chemical Name or Material (4-Pyridylthio)acetic acid
TSCA TSCA
Assay 98%
Molecular Formula C7H7NO2S
MDL Number MFCD00006424
CAS 10351-19-6
Beilstein 129738
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈260°C (dec.)
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.