missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Pyridylacetic Acid Hydrochloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC294750010

 View more versions of this product

Catalog No. AC294750010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Pyridylacetic acid hydrochloride
Glass bottle
139.0°C to 141.0°C
98%
97.5
97.5% min. (Argentometry)
(C5H4N)CH2COOH·HCl
2-4-pyridyl acetic acid hydrochloride, 2-pyridin-4-yl acetic acid hydrochloride, 4-acetic acid pyridine hydrochloride, 4-acetylpyridine hydrochloride, 4-carboxymethylpyridinium chloride, 4-pyridine acetic acid hydrochloride, 4-pyridineacetic acid hcl, 4-pyridineacetic acid hydrochloride, 4-pyridineacetic acid, hydrochloride, 4-pyridylacetic acid hydrochloride
C1=CN=CC=C1CC(=O)O.Cl
173.6
173.6
98%
Authentic
White to Cream
1g
6622-91-9
100.0
C7H7NO2·HCl
MFCD00012827
WKJRYVOTVRPAFN-UHFFFAOYSA-N
2-pyridin-4-ylacetic acid;hydrochloride
81097
Crystalline Powder, Crystals and/or Chunks

Chemical Identifiers

6622-91-9
173.6
WKJRYVOTVRPAFN-UHFFFAOYSA-N
81097
C1=CN=CC=C1CC(=O)O.Cl
C7H7NO2·HCl
MFCD00012827
2-4-pyridyl acetic acid hydrochloride, 2-pyridin-4-yl acetic acid hydrochloride, 4-acetic acid pyridine hydrochloride, 4-acetylpyridine hydrochloride, 4-carboxymethylpyridinium chloride, 4-pyridine acetic acid hydrochloride, 4-pyridineacetic acid hcl, 4-pyridineacetic acid hydrochloride, 4-pyridineacetic acid, hydrochloride, 4-pyridylacetic acid hydrochloride
2-pyridin-4-ylacetic acid;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 229-576-3

SDS Documents