Please login to your online account to display your discounted pricing

4-Pyridylacetic Acid Hydrochloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Pyridylacetic acid hydrochloride
Name Note 98%
CAS 6622-91-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H7NO2·HCl
Linear Formula (C5H4N)CH2COOH.HCl
Formula Weight 173.6
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC294750010

  SDS
thermo fisher scientific
AC294750010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder, Crystals and/or Chunks
Formula Weight 173.6
Quantity 1g
Color White to cream
EINECS Number 229-576-3
CAS Max % 100.0
Packaging Glass bottle
Synonym 4-Pyridineacetic acid
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material 4-Pyridylacetic acid hydrochloride
Name Note 98%
Assay Percent Range 98%
Molecular Formula C7H7NO2·HCl
Linear Formula (C5H4N)CH2COOH.HCl
MDL Number MFCD00012827
CAS 6622-91-9
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.