missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(4-Pyridyl)acetone, TCI America™

Manufacturer:  TCI America P20395G

 View more versions of this product

Catalog No. P20395G



Specifications

Specifications

(4-Pyridyl)acetone
Yellow
6304-16-1
MFCD00129043
ILRVKOYYFFNXDB-UHFFFAOYSA-N
1-pyridin-4-ylpropan-2-one
80552
Clear Liquid at 20°C
80°C
5g
C8H9NO
4-pyridyl acetone, 1-4-pyridinyl-2-propanone, 1-pyridin-4-yl propan-2-one, 4-pyridyl acetone, 1-4-pyridyl acetone, 4-acetonylpyridine, 4-pyridinylacetone, 4-pyridylacetone, unii-r6onm9f2au, 2-propanone, 1-4-pyridinyl
CC(=O)CC1=CC=NC=C1
135.166
135.17
≥98.0% (GC)

Chemical Identifiers

6304-16-1
135.166
ILRVKOYYFFNXDB-UHFFFAOYSA-N
80552
CC(=O)CC1=CC=NC=C1
C8H9NO
MFCD00129043
4-pyridyl acetone, 1-4-pyridinyl-2-propanone, 1-pyridin-4-yl propan-2-one, 4-pyridyl acetone, 1-4-pyridyl acetone, 4-acetonylpyridine, 4-pyridinylacetone, 4-pyridylacetone, unii-r6onm9f2au, 2-propanone, 1-4-pyridinyl
1-pyridin-4-ylpropan-2-one
Safety and Handling

Safety and Handling

TSCA : No

Documents