Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Pyridyl)acetone, 98%

$57.86 - $204.92

Chemical Identifiers

Chemical Name or Material (4-Pyridyl)acetone
CAS 6304-16-1
Molecular Formula C8H9NO
Formula Weight 135.17
MDL Number MFCD00129043
Beilstein 110960
Synonym 1-(4-Pyridyl)-2-propanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1866703

  SDS
alfa aesar™
L1866703
1g Each for $57.86

AAL1866706

  SDS
alfa aesar™
L1866706
5g Each for $204.92
Description & Specifications

Specifications

Formula Weight 135.17
Chemical Name or Material (4-Pyridyl)acetone
CAS 6304-16-1
Molecular Formula C8H9NO
Assay 98%
MDL Number MFCD00129043
Melting Point 13°C
Beilstein 110960
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Quantity 1g
Boiling Point 143°C (20mmHg)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 1-(4-Pyridyl)-2-propanone

Applications
(4-Pyridyl)acetone is a reactant in the synthesis of Milrinone (M344680), a selective phosphodiesterase inhibitor with vasodilating and positive inotropic activity. Cardiotonic.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.