Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Pyridineethanesulfonic acid, 99%

Chemical Identifiers

Chemical Name or Material 4-Pyridineethanesulfonic acid
CAS 53054-76-5
Molecular Formula C7H9NO3S
Formula Weight 187.22
MDL Number MFCD00006452
UN Number UN2585
Beilstein 136690
Synonym 2-(4-Pyridyl)ethanesulfonic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1190614

  SDS
alfa aesar™
L1190614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1190622

  SDS
alfa aesar™
L1190622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 187.22
Assay 99%
Chemical Name or Material 4-Pyridineethanesulfonic acid
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Quantity 25g
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID
TSCA TSCA
Melting Point ≈295°C
Beilstein 136690
CAS 53054-76-5
MDL Number MFCD00006452
UN Number UN2585
Synonym 2-(4-Pyridyl)ethanesulfonic acid
Molecular Formula C7H9NO3S
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.