missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Pyridineethanesulfonic acid, 99%

Manufacturer:  Alfa Aesar™ L1190614

 View more versions of this product

Catalog No. AAL1190614



Specifications

Specifications

4-Pyridineethanesulfonic acid
25g
C7H9NO3S
UN2585
4-pyridineethanesulfonic acid, 2-4-pyridyl ethanesulfonic acid, 4-pyridylethylsulfonic acid, 2-pyridin-4-yl ethanesulfonic acid, 4-pyridyl-2-ethyl sulfonic acid, 4-pyridineethanesulfonicacid, 2-4-pyridyl ethanesulphonic acid, 4-2-sulfoethyl pyridine, acmc-209l3n, 4-2-sulfoethyl-pyridine
C1=CN=CC=C1CCS(=O)(=O)O
187.213
187.22
~295°C
53054-76-5
MFCD00006452
136690
RGIIAYDCZSXHGL-UHFFFAOYSA-N
2-pyridin-4-ylethanesulfonic acid
96722
99%

Chemical Identifiers

53054-76-5
187.213
RGIIAYDCZSXHGL-UHFFFAOYSA-N
96722
C1=CN=CC=C1CCS(=O)(=O)O
C7H9NO3S
MFCD00006452
4-pyridineethanesulfonic acid, 2-4-pyridyl ethanesulfonic acid, 4-pyridylethylsulfonic acid, 2-pyridin-4-yl ethanesulfonic acid, 4-pyridyl-2-ethyl sulfonic acid, 4-pyridineethanesulfonicacid, 2-4-pyridyl ethanesulphonic acid, 4-2-sulfoethyl pyridine, acmc-209l3n, 4-2-sulfoethyl-pyridine
2-pyridin-4-ylethanesulfonic acid
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID

TSCA : TSCA

Documents