missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Pyridineacetonitrile, ≥97%

$107.18 - $340.81

Chemical Identifiers

CAS 13121-99-8
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD03412601
InChI Key BMVSAKPRNWZCPG-UHFFFAOYSA-N
Synonym pyridin-4-yl-acetonitrile, 2-pyridin-4-yl acetonitrile, 4-pyridylacetonitrile, 4-pyridineacetonitrile, pyridin-4-ylacetonitrile, pyridine-4-acetonitrile, rarechem ak ml 0204, 2-4-pyridyl acetonitrile, 4-pyridylacetonitrilehydrochloride, 4-cyanomethyl-pyridine
PubChem CID 4112085
IUPAC Name 2-pyridin-4-ylacetonitrile
SMILES C1=CN=CC=C1CC#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64588MD View Documents Alfa Aesar™
H64588MD
250mg Each for $107.18
 
AAH6458803 View Documents Alfa Aesar™
H6458803
1g Each for $340.81
 
Description

Description

Applications
A pyridine derivative used in the preparation of inhibitors of Acyl CoA, cholesterol acyltransferase as well as bone morphogenetic protein (BMP) signaling inhibitors.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

13121-99-8
118.139
BMVSAKPRNWZCPG-UHFFFAOYSA-N
4112085
C1=CN=CC=C1CC#N
C7H6N2
MFCD03412601
pyridin-4-yl-acetonitrile, 2-pyridin-4-yl acetonitrile, 4-pyridylacetonitrile, 4-pyridineacetonitrile, pyridin-4-ylacetonitrile, pyridine-4-acetonitrile, rarechem ak ml 0204, 2-4-pyridyl acetonitrile, 4-pyridylacetonitrilehydrochloride, 4-cyanomethyl-pyridine
2-pyridin-4-ylacetonitrile

Specifications

250mg
C7H6N2
UN3439
pyridin-4-yl-acetonitrile, 2-pyridin-4-yl acetonitrile, 4-pyridylacetonitrile, 4-pyridineacetonitrile, pyridin-4-ylacetonitrile, pyridine-4-acetonitrile, rarechem ak ml 0204, 2-4-pyridyl acetonitrile, 4-pyridylacetonitrilehydrochloride, 4-cyanomethyl-pyridine
C1=CN=CC=C1CC#N
118.139
118.14
≥97%
13121-99-8
MFCD03412601
471559
BMVSAKPRNWZCPG-UHFFFAOYSA-N
2-pyridin-4-ylacetonitrile
4112085
Solid
4-Pyridineacetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.