Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Pyridineacetamide, 98%

$84.82 - $1,108.97

Chemical Identifiers

Chemical Name or Material 4-Pyridineacetamide
CAS 39640-62-5
Molecular Formula C7H8N2O
Formula Weight 136.15
MDL Number MFCD02685124
Synonym 2-(4-Pyridyl)acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2473403

  SDS
alfa aesar™
B2473403
1g Each for $84.82

AAB2473406

  SDS
alfa aesar™
B2473406
5g Each for $290.81

AAB2473414

  SDS
alfa aesar™
B2473414
25g Each for $1,108.97
Description & Specifications

Specifications

Formula Weight 136.15
CAS 39640-62-5
Synonym 2-(4-Pyridyl)acetamide
Molecular Formula C7H8N2O
MDL Number MFCD02685124
Chemical Name or Material 4-Pyridineacetamide
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 144° to 147°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.