Please login to your online account to display your discounted pricing

4-Pyridin-4-ylbenzaldehyde, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC35404DA

 View more versions of this product

Catalog No. CC35404DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 183.21
Quantity 1g
Assay 97%
Chemical Name or Material 4-Pyridin-4-ylbenzaldehyde
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C12H9NO
CAS 99163-12-9

Specifications

Chemical Name or Material 4-Pyridin-4-ylbenzaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H9NO
Formula Weight 183.21