missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Pyridin-4-ylbenzaldehyde, 97%, Maybridge

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C12H9NO
Formula Weight 183.21
Chemical Name or Material 4-Pyridin-4-ylbenzaldehyde
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC35404CB SDS Fisher Scientific Company
CC35404CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
183.21
97%
C12H9NO
4-Pyridin-4-ylbenzaldehyde

Specifications

183.21
99163-12-9
97%
4-Pyridin-4-ylbenzaldehyde
250mg
C12H9NO
Amber Glass Bottle
97%