Please login to your online account to display your discounted pricing

4-(pyrid-2-yloxy)benzaldehyde, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(Pyrid-2-yloxy)benzaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H9NO2
Formula Weight 199.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC48904DE

 
fisher scientific company
CC48904DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC48904CB

  SDS
fisher scientific company
CC48904CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC48904DA

  SDS
fisher scientific company
CC48904DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 199.21
Quantity 5g
Assay 97%
CAS 194017-69-1
Chemical Name or Material 4-(Pyrid-2-yloxy)benzaldehyde
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C12H9NO2