Please login to your online account to display your discounted pricing

4-Propylbenzoyl Chloride +99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Propylbenzoyl chloride
Name Note 99+%
CAS 52710-27-7
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0% (RCOCL)
Assay Percent Range 99+%
Molecular Formula C10H11ClO
Formula Weight 182.65
MDL Number MFCD00000698
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC418440250

  SDS
thermo fisher scientific
AC418440250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0% (RCOCL)
Formula Weight 182.65
Physical Form Opalescent Liquid
Color Colorless
Quantity 25g
TSCA TSCA
Density 1.0900g/mL
CAS Min % 99.0
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.5394 to 1.5404 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 52710-27-7
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
MDL Number MFCD00000698
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material 4-Propylbenzoyl chloride
Assay Percent Range 99+%
Name Note 99+%
Molecular Formula C10H11ClO
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.