missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Propoxyphenol, 96%, ACROS Organics™

Chemical Identifiers

CAS 18979-50-5
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C9H12O2
Linear Formula CH3CH2CH2OC6H4OH
Formula Weight 152.19
MDL Number MFCD00002335
Synonym Hydroquinone monopropyl ether
Chemical Name or Material 4-Propoxyphenol, 96%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC213320500 SDS Thermo Fisher Scientific
AC213320500
Glass bottle 50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

18979-50-5
100.0
C9H12O2
152.19
Hydroquinone monopropyl ether
95.0
96%
CH3CH2CH2OC6H4OH
MFCD00002335
4-Propoxyphenol, 96%

Specifications

152.19
95.0
Hydroquinone monopropyl ether
C9H12O2
MFCD00002335
CH3CH2CH2OC6H4OH
50g
100.0
Glass bottle
96%
18979-50-5
4-Propoxyphenol, 96%